• USA Home
  • 49973 - (R)-3-(4-Cyanophenoxy)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropionamide

EMAIL THIS PAGE TO A FRIEND
49973 Supelco

(R)-3-(4-Cyanophenoxy)-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropionamide

analytical standard

  • CAS Number 1132656-73-5

  • Empirical Formula (Hill Notation) C19H14F3N3O3

  • Molecular Weight 389.33

  •  Beilstein/REAXYS Number 21172990

  •  MDL number MFCD22572923

  •  PubChem Substance ID 329757551

Purchase

Properties

Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
optical activity   [α]/D 11±1°, c = 0.1 in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Forensics and Toxicology
Pharmaceutical (small molecule)
format   neat
SMILES string   C[C@@](O)(COc1ccc(cc1)C#N)C(=O)Nc2ccc(C#N)c(c2)C(F)(F)F
InChI   1S/C19H14F3N3O3/c1-18(27,11-28-15-6-2-12(9-23)3-7-15)17(26)25-14-5-4-13(10-24)16(8-14)19(20,21)22/h2-8,27H,11H2,1H3,(H,25,26)/t18-/m1/s1
InChI key   JNGVJMBLXIUVRD-GOSISDBHSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?