• USA Home
  • 50299 - (−)-Epigallocatechin gallate

EMAIL THIS PAGE TO A FRIEND
50299 Supelco

(−)-Epigallocatechin gallate

≥97.0% (HPLC)

Synonym: (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG

  • CAS Number 989-51-5

  • Empirical Formula (Hill Notation) C22H18O11

  • Molecular Weight 458.37

  •  MDL number MFCD00075940

  •  PubChem Substance ID 329757587

Purchase

Properties

Related Categories Additional Standards, Alcohols, Analytical Standards, Analytical/Chromatography, Aromatic,
assay   ≥97.0% (HPLC)
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C
SMILES string   Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4
InChI   1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1
InChI key   WMBWREPUVVBILR-WIYYLYMNSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
KB5200000
Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

HPLC Analysis of Catechins on Ascentis® RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Catechins on Ascentis® RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Mass spectrometry, Vitamins

NF-κB and Inflammation

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer. Oxidative cellular ...
BioFiles 2007, 2.4, 14.
Keywords: Acetylations, Adhesion, Angiogenesis, Apoptosis, Cancer, Carcinogens, Catalysis, Cell culture, Cell proliferation, Cell signaling, Cellular processes, Degradations, Diabetes, Diseases, Environmental, Gene expression, Glycosylations, High performance liquid chromatography, Inflammation, PAGE, Phosphorylations, Thin layer chromatography, Transcription, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?