EMAIL THIS PAGE TO A FRIEND
50323 Supelco

Disperse Orange 37

analytical standard

Synonym: 3-[4-(2,6-Dichloro-4-nitrophenylazo)-N-ethylanilino]propionitrile

  • CAS Number 13301-61-6

  • Empirical Formula (Hill Notation) C17H15Cl2N5O2

  • Molecular Weight 392.24

  •  Beilstein/REAXYS Number 6348868

  •  EC Number 236-325-1

  •  MDL number MFCD00181820

  •  PubChem Substance ID 329757588

  •  NACRES NA.24

Purchase

Properties

Related Categories 2002/61/EC, 67/548/EEC, Additional Standards, All alphabetical sorted, Analytical Standards,
Quality Level   100
grade   analytical standard
assay   ≥96.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
format   neat
SMILES string   CCN(CCC#N)c1ccc(cc1)\N=N\c2c(Cl)cc(cc2Cl)[N+]([O-])=O
InChI   1S/C17H15Cl2N5O2/c1-2-23(9-3-8-20)13-6-4-12(5-7-13)21-22-17-15(18)10-14(24(25)26)11-16(17)19/h4-7,10-11H,2-3,9H2,1H3/b22-21+
InChI key   KHZRTXVUEZJYNE-QURGRASLSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?