• USA Home
  • 50610 - Glycolic acid solution

EMAIL THIS PAGE TO A FRIEND
50610 Sigma-Aldrich

Glycolic acid solution

technical, ~55% in H2O

Synonym: Hydroxyacetic acid

  • CAS Number 79-14-1

  • Linear Formula HOCH2COOH

  • Molecular Weight 76.05

  •  Beilstein Registry Number 1209322

  •  MDL number MFCD00004312

  •  PubChem Substance ID 329757612

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   AEMRFAOFKBGASW-UHFFFAOYSA-N
grade   technical
concentration   ~55% in H2O
impurities   ~14% NaCl
refractive index   n20/D 1.424

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?