• USA Home
  • 50930 - Guanidine carbonate salt

EMAIL THIS PAGE TO A FRIEND
50930 Sigma-Aldrich

Guanidine carbonate salt

purum, ≥97.0% (NT)

  • CAS Number 593-85-1

  • Linear Formula NH2C(=NH)NH2 · ½H2CO3

  • Molecular Weight 90.08

  •  Beilstein/REAXYS Number 3628359

  •  EC Number 209-813-7

  •  MDL number MFCD00013029

  •  PubChem Substance ID 329757651

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Guanidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
grade   purum
assay   ≥97.0% (NT)
mp   >300 °C (lit.)
SMILES string   NC(N)=N.NC(N)=N.OC(O)=O
InChI   1S/2CH5N3.CH2O3/c3*2-1(3)4/h2*(H5,2,3,4);(H2,2,3,4)
InChI key   STIAPHVBRDNOAJ-UHFFFAOYSA-N
Gene Information   human ... KCNA1(3736), KCNA10(3744), KCNA2(3737), KCNA3(3738), KCNA4(3739), KCNA5(3741), KCNA6(3742), KCNA7(3743), KCNB1(3745), KCNB2(9312), KCNC1(3746), KCNC2(3747), KCNC3(3748), KCNC4(3749), KCND1(3750), KCND2(3751), KCND3(3752), KCNF1(3754), KCNG1(3755), KCNG2(26251), KCNG3(170850), KCNG4(93107), KCNH1(3756), KCNH2(3757), KCNH3(23416), KCNH4(23415), KCNH5(27133), KCNH6(81033), KCNH7(90134), KCNH8(131096), KCNQ1(3784), KCNQ2(3785), KCNQ3(3786), KCNQ4(9132), KCNQ5(56479), KCNS1(3787), KCNS2(3788), KCNS3(3790), KCNV1(27012), KCNV2(169522)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FG1750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?