• USA Home
  • 514160 - trans-4-[4-(Dimethylamino)styryl]-1-methylpyridinium p-toluenesulfonate

EMAIL THIS PAGE TO A FRIEND
514160 Sigma-Aldrich

trans-4-[4-(Dimethylamino)styryl]-1-methylpyridinium p-toluenesulfonate

98%

  • CAS Number 80969-52-4

  • Empirical Formula (Hill Notation) C23H26N2O3S

  • Molecular Weight 410.53

  •  MDL number MFCD00801023

  •  PubChem Substance ID 24873746

Purchase

Properties

assay   98%
mp   258-259 °C (lit.)
absorption   λmax 271 nm
  λmax 475 nm (2nd)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?