• USA Home
  • 51439 - Manganese Standard for IC

EMAIL THIS PAGE TO A FRIEND
51439 Supelco

Manganese Standard for IC

TraceCERT®, 1000 mg/L Mn+2 in nitric acid

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Anions and Cations Standards, Cationic Solutions,
Quality Level   300
grade   certified reference material
  TraceCERT®
concentration   1000 mg/L Mn+2 in nitric acid
application(s)   ion chromatography: suitable
format   single component solution

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the label.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L Mn2+ in ≤0.1% nitric acid, prepared with high purity Mn metal, HNO3 and water

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

78737 Nitric acid/Rubidium nitrate concentrate, IC eluent concentrate (20x) for Metrosep C Supp 1 & 2
78348 Sodium acetate/Sodium hydroxide eluent, 10 mM CH3COONa in H2O, 100 mM NaOH in H2O, IC eluent for Metrosep Carb 2
52411 Heptafluorobutyric acid, for ion chromatography, ≥99.5% (GC)
03918 DL-Tartaric acid, anhydrous, for ion chromatography, ≥99.5% (T)
02321 2,6-Pyridinedicarboxylic acid, for ion chromatography, ≥99.5% (T)
86854 Tetrabutylammonium hydroxide solution, ~40% in water, for ion chromatography
72064 Sodium hydroxide solution, 49-51% in water, eluent for IC
55517 Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
68279 Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
36486 Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
56169 Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
68487 Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
16355 Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
50439 Perchloric acid concentrate, 0.01 M HClO4 in water (0.01N), eluent concentrate for IC
43617 Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
50972 2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), for ion chromatography, eluent concentrate
61699 Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
00612 Water, for ion chromatography
61905 Nitric acid/Dipicolinic acid concentrate, HNO3 34 mM and Dipicolinic acid 14 mM in water, IC eluent concentrate (20x) for Metrosep C 4
62414 Sodium bicarbonate/Sodium carbonate concentrate, Na2CO3 64mM and NaHCO3 20mM in water, IC eluent concentrate (20x) for Metrosep A Supp 5
72784 Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
75335 Sodium bicarbonate/Sodium carbonate concentrate, Na2CO3 100 mM and NaHCO3 100 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 10
38302 Sodium carbonate/Sodium hydroxide concentrate, Na2CO3 150 mM and NaOH 15 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 16
19399 Nitric acid/Dipicolinic acid concentrate, HNO3 17 mM and Dipicolinic acid 17 mM in water, IC eluent concentrate (10x) for Metrosep C 6
69523 Sodium bicarbonate/Sodium carbonate concentrate, Na2CO3 36 mM and NaHCO3 34 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 4

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?