EMAIL THIS PAGE TO A FRIEND
51853 Sigma-Aldrich

Cannabidiol solution

~1 mg/mL in ethanol, analytical standard, for drug analysis

Synonym: CBD

Purchase

Properties

grade   analytical standard, for drug analysis
InChI Key   QHMBSVQNZZTUGM-ZWKOTPCHSA-N
assay   ≥98.5% (HPLC)
shelf life   limited shelf life, expiry date on the label
drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   ~1 mg/mL in ethanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   corresponds to standard for H-NMR
format   single component solution
storage temp.   2-8°C

Description

Packaging

ampoule contains 1.2mL solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution
WGK Germany 
2
Flash Point(F) 
57.2 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules
08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?