EMAIL THIS PAGE TO A FRIEND
51864 Supelco

(±)-Hesperetin

analytical standard

Synonym: 3′,5,7-Trihydroxy-4′-methoxyflavanone, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-chromanone, Hesperetin

  • CAS Number 69097-99-0

  • Empirical Formula (Hill Notation) C16H14O6

  • Molecular Weight 302.28

  •  MDL number MFCD00016956

  •  PubChem Substance ID 329757758

  •  NACRES NA.24

Purchase

Properties

Related Categories Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Citrus, E - H,
Quality Level   100
grade   analytical standard
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
solubility   ethanol: 50 mg/mL, clear to almost clear, colorless to slightly yellow
Featured Industry   Food and Beverages
format   neat
storage temp.   2-8°C
SMILES string   COc1ccc(cc1O)C2CC(=O)c3c(O)cc(O)cc3O2
InChI   1S/C16H14O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-6,14,17-19H,7H2,1H3
InChI key   AIONOLUJZLIMTK-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

LC/MS Analysis of Flavonoids on Chromolith® FastGradient RP-18 endcapped

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Flavonoids on Chromolith® FastGradient RP-18 endcapped
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?