• USA Home
  • 523348 - Poly(2-vinylpyridine)

EMAIL THIS PAGE TO A FRIEND
523348 Sigma-Aldrich

Poly(2-vinylpyridine)

analytical standard, average Mw 92,000 (Typical), average Mn 78,500 (Typical)

Purchase

Properties

Related Categories Materials Science, Polymer Molecular Weight Standards, Reference/Calibration Standards More...
grade   analytical standard
form   neat
mol wt   average Mn 78,500 (Typical)
  average Mw 92,000 (Typical)
application(s)   gel permeation chromatography (GPC): suitable
format   neat

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?