• USA Home
  • 53619 - NMR insert capillary for external locking (Benzene-d6)

EMAIL THIS PAGE TO A FRIEND
53619 Sigma-Aldrich

NMR insert capillary for external locking (Benzene-d6)

reusable, ≥99.5 atom % 2H

Synonym: NMR locking tube (Benzene-d6)

Purchase

Properties

Related Categories Accessories for NMR tubes, Labware, NMR Tubes, NMR Tubes and Accessories
isotopic purity   ≥99.5 atom % 2H
diam. × L   3 mm × 75 mm

Description

Application

application examples: NMR measurement of reactive products or solutions, assay determinations etc.

Packaging

5 capillaries each

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1114 - class 3 - PG 2 - Benzene
WGK Germany 
3
Flash Point(F) 
12.2 °F
Flash Point(C) 
-11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?