EMAIL THIS PAGE TO A FRIEND
53633 Supelco

Glabridin

analytical standard

Synonym: (R)-4-(3,4-Dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b′]dipyran-3-yl)-1,3-benzenediol

  • CAS Number 59870-68-7

  • Empirical Formula (Hill Notation) C20H20O4

  • Molecular Weight 324.37

  •  Beilstein/REAXYS Number 7141956

  •  MDL number MFCD03427694

  •  PubChem Substance ID 329757962

Purchase

Properties

Related Categories Additional Standards, Alcohols and Phenols, Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   LBQIJVLKGVZRIW-ZDUSSCGKSA-N
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Food and Beverages
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
512.6 °F
Flash Point(C) 
267 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?