EMAIL THIS PAGE TO A FRIEND
53666 Supelco

1-Furfurylpyrrole

analytical standard

Purchase

Properties

Related Categories A-F, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Almond, Analytical Standards,
grade   analytical standard
vapor density   >1 (vs air)
InChI Key   BTBFUBUCCJKJOZ-UHFFFAOYSA-N
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.530-1.534
  n20/D 1.531(lit.)
bp   76-78 °C/1 mmHg(lit.)
density   1.081 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Safety & Documentation

Safety Information

RTECS 
UX9631000
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?