EMAIL THIS PAGE TO A FRIEND
537020 Sigma-Aldrich

Acetic acid

glacial, ≥99.85%

Synonym: Glacial acetic acid

  • CAS Number 64-19-7

  • Linear Formula CH3CO2H

  • Molecular Weight 60.05

  •  Beilstein Registry Number 506007

  •  EC Number 200-580-7

  •  MDL number MFCD00036152

  •  PubChem Substance ID 24878187

Purchase

Properties

vapor density   2.07 (vs air)
InChI Key   QTBSBXVTEAMEQO-UHFFFAOYSA-N
assay   ≥99.85%
autoignition temp.   800 °F
expl. lim.   16 %, 92 °F
  4 %, 59 °F
refractive index   n20/D 1.371(lit.)
bp   117-118 °C(lit.)
mp   16.2 °C (lit.)
density   1.049 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by A6283-Sigma-Aldrich | Acetic acid, ReagentPlus, ≥99% | CH3CO2H

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2789PSN1 3(8) / PGII
WGK Germany 
1
RTECS 
AF1225000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?