EMAIL THIS PAGE TO A FRIEND
537039 Sigma-Aldrich

Methyl acetate

reagent grade, 95%

Purchase

Properties

Related Categories ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads, Reagent, Reagent Grade Solvents,
grade   reagent grade
vapor density   2.55 (vs air)
vapor pressure   165 mmHg ( 20 °C)
assay   95%
autoignition temp.   936 °F
expl. lim.   16 %
impurities   ≤1.50% (water)
refractive index   n20/D 1.361 (lit.)
bp   57-58 °C (lit.)
mp   −98 °C (lit.)
density   0.934 g/mL at 25 °C
storage temp.   room temp
SMILES string   COC(C)=O
InChI   1S/C3H6O2/c1-3(4)5-2/h1-2H3
InChI key   KXKVLQRXCPHEJC-UHFFFAOYSA-N

Description

Other Notes

This product has been replaced by 186325-Sigma-Aldrich | Methyl acetate, ReagentPlus, 99% | CH3COOCH3

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Central nervous system
Supplemental Hazard Statements 
EUH066
RIDADR 
UN1231 - class 3 - PG 2 - Methyl acetate
WGK Germany 
WGK 1
RTECS 
AI9100000
Flash Point(F) 
8.6 °F - closed cup
Flash Point(C) 
-13 °C - closed cup

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?