• USA Home
  • 53835 - (S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene

EMAIL THIS PAGE TO A FRIEND
53835 Supelco

(S)-5-Allyl-2-oxabicyclo[3.3.0]oct-8-ene

for chiral derivatization, ≥97.0% (GC)

Synonym: (3aS)-3,3a,4,5-Tetrahydro-3a-(2-propenyl)-2H-cyclopenta[b]furan

  • CAS Number 1052236-86-8

  • Empirical Formula (Hill Notation) C10H14O

  • Molecular Weight 150.22

  •  MDL number MFCD09038495

  •  PubChem Substance ID 329758006

  •  NACRES NA.22

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis, Chiral,
Quality Level   100
grade   for chiral derivatization
InChI Key   WBRYUDKSKFXYOD-SNVBAGLBSA-N
assay   ≥97.0% (GC)
optical activity   [α]/D -108±5°, c = 1 in methanol
optical purity   enantiomeric ratio: ≥99.0:1.0 (GC)
refractive index   n20/D 1.497

Description

General description

Widely applicable for alcohols and thiols which have been hard to be chiral-resolved with existing methods. Enabling to reduce production costs because of easy recovery.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?