• USA Home
  • 55821 - Ethyl 3-hydroxybutyrate

EMAIL THIS PAGE TO A FRIEND
55821 Supelco

Ethyl 3-hydroxybutyrate

analytical standard

  • CAS Number 5405-41-4

  • Linear Formula CH3CH(OH)CH2COOC2H5

  • Molecular Weight 132.16

  •  Beilstein/REAXYS Number 1446190

  •  MDL number MFCD00004545

  •  PubChem Substance ID 329758160

  •  NACRES NA.24

Purchase

Properties

Related Categories A-F, A-L, Acid, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted,
Quality Level   100
grade   analytical standard
assay   ≥98.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.42 (lit.)
  n20/D 1.421
bp   170 °C (lit.)
density   1.017 g/mL at 25 °C (lit.)
Featured Industry   Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care
format   neat
SMILES string   CCOC(=O)CC(C)O
InChI   1S/C6H12O3/c1-3-9-6(8)4-5(2)7/h5,7H,3-4H2,1-2H3
InChI key   OMSUIQOIVADKIM-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
170.6 °F - closed cup
Flash Point(C) 
77 °C - closed cup
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?