• USA Home
  • 56452 - 3,5-Dihydroxyhydrocinnamic acid

EMAIL THIS PAGE TO A FRIEND
56452 Supelco

3,5-Dihydroxyhydrocinnamic acid

analytical standard

Synonym: 3-(3,5-Dihydroxyphenyl)-1-propanoic acid, 3-(3,5-Dihydroxyphenyl)propionic acid

  • CAS Number 26539-01-5

  • Empirical Formula (Hill Notation) C9H10O4

  • Molecular Weight 182.17

  •  Beilstein/REAXYS Number 2723516

  •  MDL number MFCD12923243

  •  PubChem Substance ID 329758211

  •  NACRES NA.24

Purchase

Properties

Related Categories Additional Standards, Alkylresorcinols, Analytical Standards, Analytical/Chromatography, Antioxidant Standards,
Quality Level   100
grade   analytical standard
assay   ≥95.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cleaning Products
Cosmetics
Food and Beverages
Personal Care
format   neat
SMILES string   OC(=O)CCc1cc(O)cc(O)c1
InChI   1S/C9H10O4/c10-7-3-6(1-2-9(12)13)4-8(11)5-7/h3-5,10-11H,1-2H2,(H,12,13)
InChI key   ITPFIKQWNDGDLG-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?