EMAIL THIS PAGE TO A FRIEND
56523 Supelco

Cycloolivil

analytical standard

Synonym: (1S,2S,3S)-1,2,3,4-Tetrahydro-3,7-dihydroxy-1-(4-hydroxy-3-methoxyphenyl)-6-methoxy-2,3-naphthalenedimethanol, (2S,3S,4S)-4-(4-Hydroxy-3-methoxyphenyl)-2,3-bis(hydroxymethyl)-7-methoxy-1,2,3,4-tetrahydro-2,6-naphthalenediol, Isoolivil

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Phenylpropanes, Phytochemical Standards
grade   analytical standard
InChI Key   KCIQZCNOUZCRGH-VOBQZIQPSA-N
assay   ≥95.0% (HPLC)
shelf life   limited shelf life, expiry date on the label
Featured Industry   Food and Beverages
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
UN 3077 9 / PGIII
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?