• USA Home
  • 59859 - PALCAM Listeria Selective Enrichment Broth, Vegitone

EMAIL THIS PAGE TO A FRIEND
59859 Millipore

PALCAM Listeria Selective Enrichment Broth, Vegitone

for microbiology

Synonym: Vegitone PALCAM Listeria Selective Enrichment Broth

  •  NACRES NA.85

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Liquid Media/Broths, Listeria, Media by Application,
Quality Level   100
grade   for microbiology
form   powder
shelf life   limited shelf life, expiry date on the label
composition   ammonium ferric citrate, 0.5 g/L
  esculin, 0.8 g/L
  lithium chloride, 10.0 g/L
  D-mannitol, 5.0 g/L
  peptone (vegetable), 23.0 g/L
  phenol red, 0.08 g/L
  polysorbate 80, 2.0 g/L
  soya lecithin, 1.0 g/L
  yeast extract, 5.0 g/L
application(s)   microbiological culture: suitable
  pathogen testing: suitable
  sample preparation: suitable
final pH   7.4±0.2 (25 °C)
Featured Industry   Agriculture
Environmental
Food and Beverages
Veterinary
suitability   selective and differential for Listeria spp.

Description

Application

This PALCAM Listeria Selective Enrichment Broth contains plant peptone instead of animal derived peptone. With added supplement it is recommended for selective enrichment and identification of Listeria species. It is also mentioned below the ISO specification 11290-1:1996.

Preparation Note

Supplement: 2 vial/l of PALCAM Listeria Selective Supplement according to Van Netten et al. (Cat. No. 91986)

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?