• USA Home
  • 60219 - Potassium phosphate monobasic

EMAIL THIS PAGE TO A FRIEND
60219 Sigma-Aldrich

Potassium phosphate monobasic

BioUltra, anhydrous, ≥99.5% (T)

Synonym: Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Purchase

Properties

product line   BioUltra
assay   ≥99.5% (T)
impurities   insoluble matter, passes filter test
  ≤0.001% total nitrogen (N)
pH   4.0-4.5 (25 °C, 1 M in H2O)
pKa (25 °C)   (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
mp   252.6 °C (lit.)
solubility   H2O: 1 M at 20 °C, clear, colorless
density   2.338 g/mL at 25 °C (lit.)
anion traces   chloride (Cl-): ≤5 mg/kg
  sulfate (SO42-): ≤30 mg/kg
cation traces   Al: ≤5 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤200 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: 0.046
  λ: 280 nm Amax: 0.040
SMILES string   [K+].OP(O)([O-])=O
InChI   1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1
InChI key   GNSKLFRGEWLPPA-UHFFFAOYSA-M

Description

Other Notes

This product has been replaced by 60218-SIGMA | Potassium phosphate monobasic BioUltra, for molecular biology, anhydrous, ≥99.5% (T)

Primary standard buffer; Buffer component of many culture media

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TC6615500

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?