EMAIL THIS PAGE TO A FRIEND
60870 Sigma-Aldrich

Carbon monoxide

puriss., ≥99.0%

  • CAS Number 630-08-0

  • Empirical Formula (Hill Notation) CO

  • Molecular Weight 28.01

  •  Beilstein Registry Number 3587264

  •  EC Number 211-128-3

  •  MDL number MFCD00011320

  •  PubChem Substance ID 24881997

Purchase

Properties

InChI Key   UGFAIRIUMAVXCW-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0%
autoignition temp.   1128 °F
expl. lim.   74 %
impurities   ≤0.1% H2
  ≤0.5% N2
  ≤0.5% O2+Ar
bp   −191.5 °C(lit.)
mp   −205 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Packaging

Cylinder with net ~3 kg

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1016 - class 2 - Carbon monoxide, compressed
WGK Germany 
1
RTECS 
FG3500000
Protocols & Articles
Related Products

control valve

Product #

Description

Add to Cart

99113 Outlet valve No 70690, stainless steel, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?