• USA Home
  • 610577 - (±)-Cotinine-(methyl-d3) solution

EMAIL THIS PAGE TO A FRIEND
610577 Supelco

(±)-Cotinine-(methyl-d3) solution

99 atom % D, 1 mg/mL in methanol, drug standard

Purchase

Properties

Related Categories 1694 Pharmaceuticals & Personal Care Products, Additional Standards, Alkaloids, Analytical Standards, Analytical/Chromatography,
grade   drug standard
Agency/Method   EPA 1694
isotopic purity   99 atom % D
availability   not available in Japan
concentration   1 mg/mL in methanol
Featured Industry   Environmental
Pharmaceutical (small molecule)
mass shift   M+3
storage temp.   −20°C
SMILES string   [2H]C([2H])([2H])N1C(CCC1=O)c2cccnc2
InChI   1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/i1D3
InChI key   UIKROCXWUNQSPJ-FIBGUPNXSA-N

Description

Packaging

1 mL in ampule

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Eyes
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
WGK 2
Flash Point(F) 
51.8 °F - closed cup - Not applicable
Flash Point(C) 
11 °C - closed cup - Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?