EMAIL THIS PAGE TO A FRIEND
61220 Sigma-Aldrich

Cupferron

ACS reagent, for the determination and separation of Ce, Cu, Fe, Ga, Nb, Sn, Ta, Ti, U, V, W

Synonym: N-Nitroso-N-phenylhydroxylamine ammonium salt

  • CAS Number 135-20-6

  • Linear Formula C6H5N(NO)ONH4

  • Molecular Weight 155.15

  •  Beilstein Registry Number 3919107

  •  EC Number 205-183-2

  •  MDL number MFCD00078422

Purchase

Properties

grade   ACS reagent
InChI Key   GDEBSAWXIHEMNF-UHFFFAOYSA-O
quality   for the determination and separation of Ce, Cu, Fe, Ga, Nb, Sn, Ta, Ti, U, V, W
mp   150-155 °C (dec.) (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
NC4725000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

675636 Cupferron, 97%, reagent grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?