• USA Home
  • 613347 - Fluoxetine-d5 solution

EMAIL THIS PAGE TO A FRIEND
613347 Supelco

Fluoxetine-d5 solution

1 mg/mL in methanol, 98 atom % D, drug standard

Synonym: Fluoxetine-phenyl-d5 solution

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Antidepressants Standards, Chromatography,
grade   drug standard
isotopic purity   98 atom % D
availability   not available in Japan
concentration   1 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   single component solution
storage temp.   −20°C

Description

Application

Fluoxetine-d5 solution may be used as an internal standard in the micro-high performance liquid chromatography (micro-HPLC) or liquid chromatography coupled with mass spectrometric(LC-MS)analysis of basic drugs like quetiapine, citalopram, amitriptyline, methadone and sertraline in human plasma and acidified water samples.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Fluoxetine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-DA
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?