EMAIL THIS PAGE TO A FRIEND
61748 Sigma-Aldrich

LB Broth

for microbiology

Purchase

Properties

grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   sodium chloride, 5 g/L
  tryptone, 10 g/L
  yeast extract, 5 g/L
final pH   7.2 (37 °C)

Description

Application

For the culture of E. coli K12 strains for the preparation of phage and plasmid DNA acc. to Miller (1987).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

L3022 LB Broth (Lennox), Powder microbial growth medium

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?