• USA Home
  • 62306 - Lipase from wheat germ

EMAIL THIS PAGE TO A FRIEND
62306 Sigma-Aldrich

Lipase from wheat germ

~0.1 U/mg

Synonym: Triacylglycerol acylhydrolase, Triacylglycerol lipase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Diagnostic and Analytical Enzymes,
specific activity   ~0.1 U/mg
mol wt   Mr ~42000
storage temp.   2-8°C
Gene Information   wheat ... lipase(543006)

Description

Biochem/physiol Actions

Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol acetic acid per minute at pH 7.4 and 40°C (triacetin, Cat. No. No. 90240 as substrate)

Other Notes

Characterization; Hydrolysis of N-benzyloxycarbonyl-cis-2,6-(acetoxymethyl)piperidine

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
TO9776500
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Enzymes & Proteins

Committed to developing and providing breakthrough problem-solving reagents and solutions, explore our comprehensive offering of enzymes and protein reagents and kits with the confidence that you wil...
Keywords: Cell signaling, Diagnostic, Enzyme-linked immunosorbent assay, Immunohistochemistry, Metabolism, Molecular biology, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?