EMAIL THIS PAGE TO A FRIEND
62478 Sigma-Aldrich

Lithium chloride

puriss. p.a., ACS reagent, anhydrous, ≥99.0% (AT)

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, E-L, Puriss p.a. ACS, Essential Chemicals,
grade   ACS reagent
InChI Key   KWGKDLIKAYFUFQ-UHFFFAOYSA-M
grade   puriss. p.a.
assay   ≥99.0% (AT)
impurities   ≤0.001% total nitrogen (N)
loss   ≤1% loss on drying, 110 °C
pH   5.5-7.5 (25 °C, 50 mg/mL in H2O)
mp   605 °C(lit.)
anion traces   nitrate (NO3-): ≤10 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ba: ≤10 mg/kg
  Ca: ≤20 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤10 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Lithium chloride (LiCl) may be used in the synthesis of lithium perchlorate by reacting with perchloric acid.

General description

Lithium chloride (LiCl) participates in the Horner-Wadsworth-Emmons reactions of base-sensitive aldehydes and phosphonates. LiCl/N,N-Dimethylacetamide (DMAc) solvent system has been employed for dissolution of cellulose and synthesis of various cellulose derivatives (cellulose esters, carbamates, sulphonates and ethers). It can be synthesized from lithium carbonate (Li2CO3).

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OJ5950000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?