EMAIL THIS PAGE TO A FRIEND
62610 Sigma-Aldrich

Lithium tetraborate

puriss. p.a., ACS reagent, ≥98.0% (T)

Purchase

Properties

grade   ACS reagent
  puriss. p.a.
assay   ≥98.0% (T)
  98.0-102.0% (ACS, T)
loss   2.0% loss on fusion, 950°C
mp   760-880 °C (lit.)
density   ≥0.25 g/mL at 25 °C (lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ag: ≤5 mg/kg
  Al: ≤5 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Tl: ≤10 mg/kg
  Zn: ≤5 mg/kg
Featured Industry   Forensics and Toxicology
SMILES string   [Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O
InChI   1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1
InChI key   PSHMSSXLYVAENJ-UHFFFAOYSA-N

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Reagent for the decomposition of oxides and silicates by fusion

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?