• USA Home
  • 62629 - Laurel oil from Laurus nobilis

EMAIL THIS PAGE TO A FRIEND
62629 Sigma-Aldrich

Laurel oil from Laurus nobilis

purum

Synonym: Lauri oleum

Purchase

Properties

Related Categories Alphabetical Listings, Essential Oils, Flavors and Fragrances, I-L
grade   purum
storage temp.   2-8°C

Description

Analysis Note

corresponds to analytical specification of Ph Helv V

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?