EMAIL THIS PAGE TO A FRIEND
62696 Sigma-Aldrich

Luteolin

≥99.0% (TLC)

Synonym: 3′,4′,5,7-Tetrahydroxyflavone

  • CAS Number 491-70-3

  • Empirical Formula (Hill Notation) C15H10O6

  • Molecular Weight 286.24

  •  Beilstein/REAXYS Number 292084

  •  EC Number 207-741-0

  •  MDL number MFCD00017309

  •  PubChem Substance ID 329759748

Purchase

Properties

InChI Key   IQPNAANSBPBGFQ-UHFFFAOYSA-N
assay   ≥99.0% (TLC)
mp   ~330 °C (lit.)
storage temp.   2-8°C
Gene Information   human ... CDC2(983), CDK5(1020), CDK6(1021), CYP1A2(1544), GSK3A(2931)
mouse ... Hexa(15211)
rat ... Il4(287287), Tnf(24835)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
LK9275210

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

L9283 Luteolin, ≥98% (TLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?