EMAIL THIS PAGE TO A FRIEND
63290 Sigma-Aldrich

Malonic acid

puriss., ≥99.0% (T)

Synonym: Propanedioic acid

  • CAS Number 141-82-2

  • Linear Formula CH2(COOH)2

  • Molecular Weight 104.06

  •  Beilstein Registry Number 1751370

  •  EC Number 205-503-0

  •  MDL number MFCD00002707

Purchase

Properties

InChI Key   OFOBLEOULBTSOW-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (T)
ign. residue   ≤0.05%
mp   132-135 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OO0175000
Flash Point(F) 
341.6 °F
Flash Point(C) 
172 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?