EMAIL THIS PAGE TO A FRIEND
63462 Sigma-Aldrich

L-(+)-Mandelic acid

purum, ≥98.0% (T)

Synonym: (S)-α-Hydroxyphenylacetic acid

  • CAS Number 17199-29-0

  • Empirical Formula (Hill Notation) C8H8O3

  • Molecular Weight 152.15

  •  Beilstein/REAXYS Number 2208678

  •  EC Number 241-240-8

  •  MDL number MFCD00004495

  •  PubChem Substance ID 329759865

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   IWYDHOAUDWTVEP-ZETCQYMHSA-N
grade   purum
assay   ≥98.0% (T)
optical activity   [α]20/D +153±5°, c = 5% in H2O
optical purity   enantiomeric ratio: ≥98:1 (HPLC)
mp   130-133 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Mandelic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?