EMAIL THIS PAGE TO A FRIEND
65372 Sigma-Aldrich

O-Methylserotonin

purum, ≥98.0% (NT)

Synonym: 2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-Methoxytryptamine, Mexamine

  • CAS Number 608-07-1

  • Empirical Formula (Hill Notation) C11H14N2O

  • Molecular Weight 190.24

  •  Beilstein/REAXYS Number 145587

  •  EC Number 210-153-7

  •  MDL number MFCD00005662

  •  PubChem Substance ID 329760061

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   JTEJPPKMYBDEMY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (NT)
mp   120-122 °C
Gene Information   human ... HTR1A(3350), HTR2A(3356), HTR2C(3358)
rat ... Htr2a(29595), Htr2c(25187), Htr7(65032)

Description

Biochem/physiol Actions

Nonselective serotonin receptor agonist that lacks affinity for the 5-HT3 receptor.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NL4059000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?