• USA Home
  • 67904 - Arsenic Standard for ICP

EMAIL THIS PAGE TO A FRIEND
67904 Supelco

Arsenic Standard for ICP

TraceCERT®, 10,000 mg/L As in nitric acid

  •  NACRES NA.24

Purchase

Properties

Quality Level   300
grade   certified reference material
  TraceCERT®
concentration   10,000 mg/L As in nitric acid
application(s)   AAS: suitable
  ICP: suitable
format   single component solution

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the label. Information about trace impurities are stated on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Preparation Note

10′000 mg/L As in 5% nitric acid, prepared with high purity As2O3, HNO3 and water

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
EUH071
RIDADR 
UN 2031B 8 / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?