• USA Home
  • 69490 - Methyltriphenoxyphosphonium iodide

EMAIL THIS PAGE TO A FRIEND
69490 Sigma-Aldrich

Methyltriphenoxyphosphonium iodide

technical, ≥96.0% (AT)

Synonym: Triphenyl phosphite methoiodide

Purchase

Properties

Related Categories C-X Bond Formation (Halogen), Chemical Synthesis, Iodination, Phase Transfer Catalysts, Phosphonium Salts,
InChI Key   VKTOBGBZBCELGC-UHFFFAOYSA-M
grade   technical
assay   ≥96.0% (AT)
mp   142-146 °C(lit.)

Description

Caution

may discolor to brown on storage

Other Notes

Reagent for the selective dehydration of secondary alcohols; Selective conversion of prim. alcohols to iodides; Deoxygenation of epoxides to olefins

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

226432 Methyltriphenoxyphosphonium iodide, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?