• USA Home
  • 69791 - β,β,3-Trimethylbenzenepropanol

EMAIL THIS PAGE TO A FRIEND
69791 Supelco

β,β,3-Trimethylbenzenepropanol

analytical standard

Synonym: 2,2-Dimethyl-3-(3-methylphenyl)propanol, 2,2-Dimethyl-3-(3-tolyl)propan-1-ol, 3-(2,2-Dimethyl-3-hydroxypropyl)toluene

  • CAS Number 103694-68-4

  • Empirical Formula (Hill Notation) C12H18O

  • Molecular Weight 178.27

  •  Beilstein/REAXYS Number 3237799

  •  MDL number MFCD03701092

  •  PubChem Substance ID 329761966

Purchase

Properties

Related Categories All Alphabetically Sorted, Allergen Standards, Analytical Standards, Analytical/Chromatography, Cosmetics & Perfumes,
grade   analytical standard
InChI Key   FYMOBFDUZIDKMI-UHFFFAOYSA-N
assay   ≥98.0% (GC)
shelf life   limited shelf life, expiry date on the label
refractive index   n20/D 1.516-1.520
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
storage temp.   2-8°C
Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
269.6 °F
Flash Point(C) 
132 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?