• USA Home
  • 70951 - Peptone from gelatin, enzymatic digest

EMAIL THIS PAGE TO A FRIEND
70951 Millipore

Peptone from gelatin, enzymatic digest

for biotechnological purposes

Synonym: Peptone from gelatin

Purchase

Properties

Related Categories Base Ingredients for Media, Microbiology, Protein Sources (Peptones & Extracts), Protein Sources (derived from animal tissue), Protein Sources (milk derived),
Quality Level   100
grade   for biotechnological purposes
assay   ≥15% total nitrogen (N) basis
form   powder
application(s)   microbiology: suitable (preparation)
impurities   ≥2.4% amino N
ign. residue   <9.0%
loss   ≤6.0% loss on drying
pH   6.5-7.5 (25 °C)
solubility   H2O: 2%, clear, yellow to brownish-yellow
Featured Industry   Food and Beverages

Description

General description

Peptone from gelatin, enzymatic digest is a rich source of amino acids and peptides.

Application

Peptone from gelatin, enzymatic digest is recommended for culture media in analytical microbiology and in industrial fermentation.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology's Basic Ingredient: Protein Sources (Peptones)

Peptones and extracts are excellent natural sources of amino acids, peptides and proteins in growth media. They are most often obtained by enzymatic digestion or acid hydrolysis of natural products, ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?