EMAIL THIS PAGE TO A FRIEND
71012BC Millipore

GDS Cronobacter Tq

BioControl, Molecular based PCR test for the detection of Cronobacter spp. in infant nutritional formulas and environmental samples

Purchase

Properties

shelf life   6‑12 mo.
packaging   pkg of 100 tests
mfr. no.   Assurance GDS®
  BioControl Systems, 71012-100
application(s)   PCR: suitable
Featured Industry   Food and Beverages
storage temp.   2-8°C
suitability   Cronobacter spp.

Description

General description

GDS for Cronobacter Tq is an automated nucleic acid amplification system for the detection of Cronobacter in infant nutritional formula, ingredients, and environmental samples.

Features and Benefits

• Simple 1-step enrichment procedure for powdered infant formula, ingredients, and environmental samples
• Results are available in just 26 hours, including enrichment, sample prep and analysis

Components

• Cronobacter Amplification Tubes Tq
• Resuspension Buffer
• PD Buffer

Compatibility

BioControl GDS

Testing Method

PCR

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Assurance® GDS CronobacterTq II Assay

Qualitative molecular method for the detection of Cronobacter spp. in infant formula, infant cereals and environmental samples.
Keywords: Environmental, Genetic, Microbiology, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?