• USA Home
  • 71906 - Sodium phosphate tribasic hexahydrate

EMAIL THIS PAGE TO A FRIEND
71906 Sigma-Aldrich

Sodium phosphate tribasic hexahydrate

purum p.a., ≥97.0% (T)

Synonym: Trisodium phosphate hexahydrate

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
InChI Key   BPFZWBABAJEKEO-UHFFFAOYSA-K
grade   purum p.a.
assay   ≥97.0% (T)
anion traces   chloride (Cl-): ≤50 mg/kg
  fluoride (F-): ≤50 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3262 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?