• USA Home
  • 72580 - Nitrite on polymer support

EMAIL THIS PAGE TO A FRIEND
72580 Sigma-Aldrich

Nitrite on polymer support

extent of labeling: ~4 mmol/g NO2 loading

Synonym: Amberlite® IRA 900 NO2--form

Purchase

Properties

Related Categories Chemical Synthesis, Other Supported Reagents, Solid Supported Synthesis, Specialty Synthesis, Supported Reagents More...
extent of labeling   ~4 mmol/g NO2 loading
impurities   ~14% water

Description

Other Notes

Polymer supported reagent for the synthesis of nitroalkanes from alkyl halides

Legal Information

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?