EMAIL THIS PAGE TO A FRIEND
73867 Supelco

3-Nonanone

analytical standard

Synonym: Ethyl hexyl ketone

  • CAS Number 925-78-0

  • Linear Formula CH3(CH2)5COC2H5

  • Molecular Weight 142.24

  •  Beilstein Registry Number 1700447

  •  EC Number 213-125-2

  •  MDL number MFCD00009541

  •  PubChem Substance ID 329765294

Purchase

Properties

Related Categories All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, Animal Products, Banana,
grade   analytical standard
InChI Key   IYTXKIXETAELAV-UHFFFAOYSA-N
assay   ≥98.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤1.0% water
refractive index   n20/D 1.42(lit.)
  n20/D 1.420-1.422
bp   187-188 °C(lit.)
density   0.821 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
UN 1993 / PGIII
WGK Germany 
2
RTECS 
RA8227000
Flash Point(F) 
154.4 °F
Flash Point(C) 
68 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?