EMAIL THIS PAGE TO A FRIEND
74001 Supelco

Dibutyl sulfide

analytical standard

Synonym: 1,1′-Thiobisbutane, Butyl sulfide, NSC 8460

  • CAS Number 544-40-1

  • Linear Formula CH3(CH2)3S(CH2)3CH3

  • Molecular Weight 146.29

  •  Beilstein Registry Number 1732829

  •  EC Number 208-870-5

  •  MDL number MFCD00009468

  •  PubChem Substance ID 329764944

Purchase

Properties

Related Categories A-L, Analytical Standards, Analytical/Chromatography, Flavor and Fragrance Standards, Food and Cosmetic Component Standards More...
grade   analytical standard
vapor density   5.07 (vs air)
vapor pressure   5.17 mmHg ( 37.7 °C)
InChI Key   HTIRHQRTDBPHNZ-UHFFFAOYSA-N
assay   ≥98.5% (GC)
shelf life   limited shelf life, expiry date on the label
impurities   ≤0.3% water
refractive index   n20/D 1.452(lit.)
  n20/D 1.452-1.454
bp   188-189 °C(lit.)
mp   −76 °C (lit.)
density   0.838 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Safety & Documentation

Safety Information

RIDADR 
UN 3334
RTECS 
ER6417000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?