EMAIL THIS PAGE TO A FRIEND
74150 Supelco

N.N.C.D.-reagent

for spectrophotometric det. of 1,2-dihydroxy benzene derivatives, ≥97.0%

Synonym: 2-Chloro-4-nitro-1-benzenediazonium 2-naphthalenesulfonate

  • CAS Number 6035-19-4

  • Empirical Formula (Hill Notation) C16H10ClN3O5S

  • Molecular Weight 391.79

  •  Beilstein/REAXYS Number 3871383

  •  EC Number 227-912-3

  •  MDL number MFCD00042809

  •  PubChem Substance ID 329765505

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
InChI Key   LKBPUFYOZCBOIB-UHFFFAOYSA-M
assay   ≥97.0% (N)
  ≥97.0%
quality   for spectrophotometric det. of 1,2-dihydroxy benzene derivatives
ign. residue   ≤0.1%
mp   146-150 °C (dec.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?