• USA Home
  • 74187 - 5-Methyl-2-thiophenecarboxaldehyde

EMAIL THIS PAGE TO A FRIEND
74187 Supelco

5-Methyl-2-thiophenecarboxaldehyde

analytical standard

Synonym: NSC 87542

  • CAS Number 13679-70-4

  • Empirical Formula (Hill Notation) C6H6OS

  • Molecular Weight 126.18

  •  Beilstein Registry Number 106896

  •  EC Number 237-178-6

  •  MDL number MFCD00005434

  •  PubChem Substance ID 329765537

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Flavor and Fragrance Standards, Food and Cosmetic Component Standards, M-Z More...
grade   analytical standard
InChI Key   VAUMDUIUEPIGHM-UHFFFAOYSA-N
assay   ≥97.0% (GC)
shelf life   limited shelf life, expiry date on the label
impurities   ≤1.0% water
refractive index   n20/D 1.580-1.586
  n20/D 1.583(lit.)
bp   114 °C/25 mmHg(lit.)
density   1.17 g/mL at 25 °C(lit.)
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
storage temp.   2-8°C

Description

General description

5-Methyl-2-thiophenecarboxaldehyde can be obtained as a product from 2-thiophenecarboxaldehyde via metalation, and alkylation, followed by a series of reactions.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?