EMAIL THIS PAGE TO A FRIEND
74191 Sigma-Aldrich

Obacunone

analytical standard

  • CAS Number 751-03-1

  • Empirical Formula (Hill Notation) C26H30O7

  • Molecular Weight 454.51

  •  MDL number MFCD00075888

  •  PubChem Substance ID 329765539

Purchase

Properties

Related Categories Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Citrus, Isoprenoids,
grade   analytical standard
InChI Key   MAYJEFRPIKEYBL-OASIGRBWSA-N
assay   ≥90.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤5.0% water
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RR1104200
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?