EMAIL THIS PAGE TO A FRIEND
74440 Sigma-Aldrich

(−)-Nopol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-ethanol, 6,6-Dimethyl-2-norpinene-2-ethanol

  • CAS Number 35836-73-8

  • Empirical Formula (Hill Notation) C11H18O

  • Molecular Weight 166.26

  •  Beilstein Registry Number 3196379

  •  EC Number 252-744-2

  •  MDL number MFCD00075187

  •  PubChem Substance ID 329765690

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   ROKSAUSPJGWCSM-UWVGGRQHSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −37.0±1°, neat
refractive index   n20/D 1.493
bp   235-236 °C(lit.)
density   0.967 g/mL at 20 °C(lit.)

Description

Other Notes

Reagent for preparing NB-enantride, a chiral trialkylborohydride for asymmetric reduction of ketones

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

340367 (1R)-(−)-Nopol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?