• USA Home
  • 75606 - (Z)-2-Methylglutaconic acid

EMAIL THIS PAGE TO A FRIEND
75606 Supelco

(Z)-2-Methylglutaconic acid

≥97.0% (HPLC)

Synonym: (Z)-2-Methyl-2-pentenedioic acid

Purchase

Properties

Quality Level   100
assay   ≥97.0% (HPLC)
mp   123-126 °C
storage temp.   2-8°C
SMILES string   OC(C/C=C(C)\C(O)=O)=O
InChI   1S/C6H8O4/c1-4(6(9)10)2-3-5(7)8/h2H,3H2,1H3,(H,7,8)(H,9,10)/b4-2-
InChI key   JKGHDBJDBRBRNA-RQOWECAXSA-N

Description

Biochem/physiol Actions

2-Methylglutaconic acid is found in the urine of patients with organic aciduria from 2-methylacetoacetyl-CoA thiolase.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?