• USA Home
  • 76381 - 4-Isothiocyanato-TEMPO

EMAIL THIS PAGE TO A FRIEND
76381 Supelco

4-Isothiocyanato-TEMPO

for ESR-spectroscopy, ≥97.0%

Synonym: 4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl

  • CAS Number 36410-81-8

  • Empirical Formula (Hill Notation) C10H17N2OS

  • Molecular Weight 213.32

  •  Beilstein/REAXYS Number 4421977

  •  MDL number MFCD00010109

  •  PubChem Substance ID 329767078

  •  NACRES NA.22

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents HPLC,
Quality Level   100
grade   for ESR-spectroscopy
assay   ≥97.0% (CHN)
  ≥97.0%
mp   123-128 °C
storage temp.   2-8°C
SMILES string   CC1(C)CC(CC(C)(C)N1[O])N=C=S
InChI   1S/C10H17N2OS/c1-9(2)5-8(11-7-14)6-10(3,4)12(9)13/h8H,5-6H2,1-4H3
InChI key   BKENZAFIBRCMKD-UHFFFAOYSA-N

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Synthesis of spin-labeled derivatives

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?