EMAIL THIS PAGE TO A FRIEND
76878 Sigma-Aldrich

Pentane

puriss., absolute, over molecular sieve (H2O ≤0.005%), ≥99.0% (GC)

  • CAS Number 109-66-0

  • Linear Formula CH3(CH2)3CH3

  • Molecular Weight 72.15

  •  Beilstein/REAXYS Number 969132

  •  EC Number 203-692-4

  •  MDL number MFCD00009498

  •  PubChem Substance ID 57652664

Purchase

Properties

vapor density   2.48 (vs air)
vapor pressure   26.98 psi
grade   absolute
  puriss.
assay   ≥99.0% (GC)
autoignition temp.   500 °F
quality   over molecular sieve (H2O ≤0.005%)
expl. lim.   ~8.3 %
refractive index   n20/D 1.358 (lit.)
  n20/D 1.358
bp   35-36 °C (lit.)
mp   −130 °C (lit.)
solubility   ethanol: soluble(lit.)
density   0.626 g/mL at 25 °C (lit.)
SMILES string   CCCCC
InChI   1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
InChI key   OFBQJSOFQDEBGM-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes
WGK Germany 
2
RTECS 
RZ9450000
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?